Learn More
Manutan Storage tray, Gratnells, red, A4 deep tray
Product Code. 12885526
Encompass_Preferred
Product Code. 12885526
Brand: Manutan A367150
Only null left
Add to basket
Add to basket
The ever popular Gratnell trays suitable for educational, office and workplace use. Made from extra strong polypropylene, resistant to the majority of chemicals. Flat bottom for easy cleaning. Shallow and jumbo trays have unique cut-out handle with stack and nest facility.
  • A4 deep type
  • Red
  • Dimension (H x W x D): 150mm x 312mm x 427mm