Learn More
BRAND™ Straight Burette Stopcock with Spare Key
Stopcock housing with precision Boro 3.3 burette tip. Interchangeable PTFE key with retention device.
Brand:  BRAND™ 81939
48.20 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Product Code. 10682052
  • 55.60 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Amber color
  • Straight stopcock for burette cap
  • Includes PTFE key
  • Stopcock size 3NS/12
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Amber
Burette
1 Each
Stopcock
50 mL
Borosilicate Glass, PTFE
Burette Cap