Learn More
Lenz Laborglasinstrumente™ Splash Heads Acc. to Stutzer, Straight
For placement onto distillation flasks for separation of drops. Lenz Laborglasinstrumente™ Splash Heads Acc. to Stutzer, Straight, are manufactured from durable Duran™ glass tubing.
Brand:  Lenz Laborglasinstrumente™ 05035045
 View more versions of this product
Product Code. 11953461
  • 104.00 € / Each
Explore more special offers
Description
Description
  • Straight with NS cone and socket
  • Temperature and chemical resistance
  • Minimal thermal expansion
Specifications
Specifications
For separation of drops (Direct placement onto distillation flasks)
Straight Splash Heads
Duran Glass
NS 29/32, NS 14/23