Learn More
Sampling Systems™ Straight Tweezers
These tweezers made of 316L stainless steel are designed for use in laboratory, production and industrial environments. Tweezers make it easy to pick up intricate items, such as vials and tissues.
Brand:  Sampling Systems™ A596-300
 View more versions of this product
Product Code. 17912112
  • 34.20 € / Each
Explore more special offers
Description
Description
  • Range of different convenient lengths
Specifications
Specifications
Straight
Stainless Steel (316L)
Tweezers
Non-sterile
11.8 in.
300 mm
1 Tweezer/Cs.
Standard