Learn More
Thermo Scientific Chemicals Straight Wall Inconel Crucible
62.41 € - 352.00 €
Specifications
Description Straight Wall
Material Inconel
Product Type Specs Inconel Crucible
View More Specs
Products 12
Product Code Brand Capacity (Metric) Diameter (Metric) Outer Price Quantity & Availability  
Product Code Brand Capacity (Metric) Diameter (Metric) Outer Price Quantity & Availability  
15466135
View Documents
Thermo Scientific Chemicals
035890.KT
5 mL Outside: 21mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15486135
View Documents
Thermo Scientific Chemicals
10 mL Outside: 27mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15406145
View Documents
Thermo Scientific Chemicals
035932.KT
15 mL Outside: 33mm
91.15 €
Each
In Stock
to see stock.
Add to basket
15426145
View Documents
Thermo Scientific Chemicals
035953.KT
20 mL Outside: 33mm
81.71 €
Each
In Stock
to see stock.
Add to basket
15446145
View Documents
Thermo Scientific Chemicals
25 mL Outside: 45mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15476145
View Documents
Thermo Scientific Chemicals
036012.KT
55 mL Outside: 46mm
94.97 €
Each
In Stock
to see stock.
Add to basket
15423625
View Documents
Thermo Scientific Chemicals
036033.KT
1000 mL Outside: 127mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15496145
View Documents
Thermo Scientific Chemicals
100 mL Outside: 59mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15426155
View Documents
Thermo Scientific Chemicals
036072.KT
55 mL Outside: 47mm
100.10 €
Each
In Stock
to see stock.
Add to basket
15473635
View Documents
Thermo Scientific Chemicals
036090.KT
500 mL Outside: 101mm
223.11 €
Each
In Stock
to see stock.
Add to basket
15456155
View Documents
Thermo Scientific Chemicals
036099.KT
75 mL Outside: 51mm
118.59 €
Each
In Stock
to see stock.
Add to basket
15461437
View Documents
Thermo Scientific Chemicals
036102.KT
250 mL Outside: 82mm
123.62 €
Each
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Straight Wall
Inconel
Inconel Crucible