missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences Strep-Tactin™ 4Flow™ high capacity 50% suspension

Strep-Tactin™ 4Flow™ high capacity 50% Suspension to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins

167.00 € - 22575.00 €

Specifications

Binding Capacity 20 mg/mL resin
Concentration 50% suspension
Description 4% agarose, crosslinked
Particle Size 50 to 150 μm
Form Resin Slurry
View More Specs

Products 4
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
30117602
View Documents
IBA Lifesciences
2-1250-002
4 mL
167.00 €
4mL
Please to purchase this item. Need a web account? Register with us today!
30117603
View Documents
IBA Lifesciences
2-1250-010
20 mL
595.00 €
20mL
Please to purchase this item. Need a web account? Register with us today!
30117610
View Documents
IBA Lifesciences
2-1250-025
50 mL
1375.00 €
50mL
Please to purchase this item. Need a web account? Register with us today!
30117606
Vedere i documenti
IBA Lifesciences
2-1250-500
1000 mL
22575.00 €
1000mL
Please to purchase this item. Need a web account? Register with us today!
Descrizione

Descrizione

  • Strep-Tactin™ 4Flow™ high capacity is a resin for purification of strep-tagged proteins via affinity chromatography
  • Product is optimized for column-based use and suitable for HPLC/FPLC
  • Recommended for all protein classes due to the broad pH compatibility, excellent pressure stability, outstanding target purity (>95%), and high binding affinity
  • 4% crosslinked agarose beads coupled to Strep-Tactin™ for straightforward purification of proteins with Strep-tag™II or a Twin-Strep-tag™
  • Targeted elution: no tedious titration steps required and multiple regeneration cycles possible
  • Ideal alternative for proteins that are difficult to elute from Strep-Tactin™XT 4Flow™ resins
  • Biotinylated proteins also bind to Strep-Tactin™ 4Flow™ high capacity and need to be blocked using BioLock
Specifica

Specifica

20 mg/mL resin
4% agarose, crosslinked
Resin Slurry
Proteins tagged with Twin-Strep-tag™; Strep-tag™II; cross reactivity: biotin
Affinity chromatography equipment
Agarose
Suspension
50% suspension
50 to 150 μm
Tetrameric protein
Bulk Resin
Twin-Strep-tag™; Strep-tag™II; cross reactivity: biotin
Bottle
Videos
SDS
Documenti

Documenti

Product Certifications
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto
IBA Lifesciences Strep-Tactin™ 4Flow™ high capacity 50% suspension

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato