Learn More
IBA Lifesciences Strep-Tactin™ Sepharose™ Resin
Strep-Tactin™ Sepharose™ resin is a 50% suspension to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
134.00 € - 15975.00 €
Specifications
Matrix Strep-Tactin Sepharose
View More Specs
Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12723074
View Documents
IBA Lifesciences
2-1201-002
4 mL
134.00 €
Each
12846436
View Documents
IBA Lifesciences
2-1201-010
20 mL N/A N/A N/A
10049889
View Documents
IBA Lifesciences
2-1201-025
50 mL N/A N/A N/A
12876436
View Documents
IBA Lifesciences
2-1201-500
1000 mL N/A N/A N/A
Description
Description
  • Strep-Tactin™ Sepharose™ is a 4% agarose coupled with the streptavidin variant Strep-Tactin™ and applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins.
  • Due to the low concentrated agarose, it is particularly suitable for large proteins, especially if the affinity to Strep-Tactin™XT 4Flow™ is too high.
  • The wide-meshed polymer allows for the simple attachment of larger proteins to the stationary phase.
  • The suspension can be used for packing of gravity flow columns since the resin is optimized for column affinity chromatography, rather than for batch purification.
  • To pack a column with 1 ml resin bed volume, 2 ml 50% suspension is necessary.
Please note that Strep-Tactin™ Sepharose™ is not compatible with HPLC/FPLC due to the low pressure stability.
Specifications
Specifications
Strep-Tactin Sepharose
Videos
SDS
Documents
Documents
Product Certifications
For Research Use Only.
Product Content Correction
Product Title
IBA Lifesciences Strep-Tactin™ Sepharose™ Resin