missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences Strep-Tactin™ Superflow™, 50% Suspension
Used for purification of recombinant Strep-tag proteins under gravity flow and low pressure or FPLC
Brand: IBA Lifesciences 2-1206-002
This item is not returnable.
View return policy
Description
- High capacity resin provides 2 to 3 times higher binding capacity for strep-tag proteins than standard Strep-Tactin Superflow suspension
- Higher yields of highly pure Strep-tag protein
- Bulk resin is recommended for packing of columns and cartridges since the resin is optimized for column affinity chromatography rather than for batch purification
- Binding capacity: 1mL sedimented resin (corresponding to 2mL of 50 % suspension) is useful for one-step purification of 50 to 100nmol recombinant Strep-tag protein (up to 3mg in case of 30 kDa protein)
Specifications
4 mL | |
2°C to 8°C | |
50% suspension |
6 months after shipping. | |
Biotin: >300 nmol/ mL resin |
Manual Strep-Tactin-Purification 2-1206-002
File size: 1.4 MB
Short Protocol Strep-Tactin-Purification 2-1206-002
File size: 695 kB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IBA Lifesciences Strep-Tactin™ Superflow™, 50% Suspension > Capacity: 4mL
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction