Learn More
IBA Lifesciences Strep-Tactin™ TACS Agarose Column
Gravity flow column suitable for traceless affinity cell and exosome isolation
Brand:  IBA Lifesciences 6-6310-001
 View more versions of this product
Product Code. 17227750
  • 111.00 € / Each
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Strep-Tactin™ TACS Agarose columns contain agarose macrobeads coated with the streptavidin variant Strep-Tactin™.
  • A special crosslinking permits the use for cell isolation and depletion approaches.
  • They were developed for column-based Traceless Affinty Cell Selection (Fab-TACS™/Nano-TACS™).
  • In this approach, Fab fragments fused to a Twin-Strep-tag™ (Fab-Streps) or nanobodies (Nano-Streps) are bound to Strep-Tactin™ on the agarose matrix.
  • After specifically capturing cells, Fab-/Nano-Streps are released from the Strep-Tactin™ Agarose by biotin addition.
  • This way the cells are eluted ready for further downstream applications.
  • The same principle is applicable for exosome isolation (Fab-TACS™ for exosomes).
  • Alternatively to Fab-Streps/Nano-Streps, it is possible to load columns with Twin-Strep-tagged, desthiobiotinylated or biotinylated antibodies (irreversible binding).
  • This makes Strep-Tactin™ TACS Agarose columns also suitable for negative cell selection or depletion approaches.
Specifications
Specifications
1 mL
Pre-packed in buffer, pH 8: 100 mM Tris-HCl pH 8.0, 1 mM EDTA, 150 mM NaCl, 0.01% sodium azide
Cross-linked agarose
Affinity Chromatographic Cell or Exosome Isolation