Learn More
IBA Lifesciences Strep-Tactin™XT 4Flow™ High Capacity Resin
BIOPHARMA
Strep-Tactin™XT 4Flow™ high capacity resin is a 50% suspension to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
Brand:  IBA Lifesciences 2-5030-500
 View more versions of this product
Product Code. 17277810
  • 22575.00 € / 1000mL
Estimated Shipment: 31-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Strep-Tactin™XT 4Flow™ is a 4% agarose coupled with the streptavidin variant Strep-Tactin™XT and applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins.
  • Due to the broad pH compatibility, excellent pressure stability, outstanding target purity (> 95%), and highest binding affinity, the resin is recommended for all protein classes, especially for low abundant or challenging proteins.
  • The low concentrated agarose allows for the simple attachment of larger proteins to the stationary phase, making it particularly suitable for purification of high-molecular-weight targets.
  • In addition, Strep-Tactin™XT 4Flow™ enables the application of denaturing agents such as urea.
  • The suspension can be used for packing of gravity flow columns since the resin is optimized for column affinity chromatography, rather than for batch purification.
  • To pack a column with 1 mL resin bed volume, 2 mL 50% suspension are necessary.
Specifications
Specifications
High capacity resin
1000 mL
Purify Strep-Tag™II or Twin-Strep-Tag™ Fusion Proteins