Learn More
IBA Lifesciences StrepMAB-Classic Conjugate
StrepMAB-Classic conjugates are monoclonal antibodies with different conjugates to detect Strep-tag™II and Twin-Strep-tag™ fusion proteins
306.00 € - 399.00 €
Specifications
Content And Storage | Undiluted: 2°C to 8°C, protect from light; Diluted: -25°C to -15°C |
---|---|
Target | Strep-tag™II and Twin-Strep-tag™ |
Type | Conjugate |
Product Code | Brand | Label or Dye | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Label or Dye | Price | Quantity & Availability | |||||
12618257
|
IBA Lifesciences
2-1509-001 |
HRP Conjugate |
398.00 €
Each |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||||
16411475
|
IBA Lifesciences
2-1563-050 |
DY-488 |
331.00 €
50µg |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||||
16441475
|
IBA Lifesciences
2-1566-050 |
DY-549 |
306.00 €
50µg |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||||
16471475
|
IBA Lifesciences
2-1569-050 |
DY-649 |
399.00 €
50µg |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||||
Description
StrepMAB-Classic is a highly selective, murine monoclonal antibody for detection of Strep-tag™II and Twin-Strep-tag™ fusion proteins in applications like western blots, ELISA, FACS, immunohistochemistry, and immunocytochemistry.It is available coupled to fluorescent dyes (DY-488, DY-549, DY-649).
The absorption/emission maxima of the fluorescent dyes are:
• DY-488: 493/518 nm
• DY-549: 550-560/575 nm
• DY-649: 655/676 nm
Please note: For capturing of Strep-tag™II and Twin-Strep-tag™ fusion proteins on solid phases, the use of StrepMAB-Immo or Strep-Tactin™XT is recommended due to the binding affinity in the pM range.
Specifications
Undiluted: 2°C to 8°C, protect from light; Diluted: -25°C to -15°C | |
Strep-tag™II and Twin-Strep-tag™ | |
Conjugate |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.