Learn More
Invitrogen™ Streptavidin Agarose, sedimented bead suspension
The streptavidin conjugated to 4% beaded crosslinked agarose can be used to isolate biotinylated proteins.
Brand: Invitrogen™ S951
376.01 EUR valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
The streptavidin conjugated to 4% beaded crosslinked agarose can be used to isolate biotinylated proteins. Additionally, biotinylated antibodies can be bound to streptavidin agarose to generate affinity matrices for the large-scale isolation of antigens.
Specifications
Streptavidin Agarose, 5mL, Affinity Column Type, Agarose Label, Suspension, Streptavidin Immobilized Phase, 2 to 8°C Storage Temperature, Wet Ice | |
5mL, 5 mL | |
Wet Ice | |
Protein assays and analysis, protein purification and isolation | |
Agarose | |
Streptavidin |
Streptavidin Agarose | |
Store in refrigerator (2°C to 8°C)., 2 to 8°C | |
Liquid Suspension | |
Agarose | |
Streptavidin Agarose | |
Affinity |
For Research Use Only. Not for use in diagnostic procedures.