missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Streptavidin, Alkaline Phosphatase Conjugate
Thermo Scientific Streptavidin, Alkaline Phosphatase Conjugate includes streptavidin in a purified form, conjugated to alkaline phosphatase for substrate based detection.
Brand: Molecular Probes™ S921
495.52 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
This item is not returnable.
View return policy
Description
Thermo Scientific Streptavidin, Alkaline Phosphatase Conjugate includes streptavidin in a purified form, conjugated to alkaline phosphatase for substrate based detection.Streptavidin is a biotin-binding protein originally isolated from Streptomyces avidinii. In contrast to avidin, streptavidin has no carbohydrate and has a mildly acidic pI of 5. Thermo Scientific streptavidin products use a recombinant form of streptavidin having a mass of 53,000 daltons and a near-neutral pI. Streptavidin is a tetrameric protein, with each subunit binding one molecule of biotin with affinity similar to that of avidin. Guanidinium chloride will dissociate avidin and streptavidin into subunits, but streptavidin is more resistant to dissociation.
Streptavidin, Alkaline Phosphatase Conjugate enables detection of biotinylated antibodies and other probes in a variety of standard assay methods, including western blotting, ELISA, immunohistochemistry, and fluorescence imaging.

Specifications
2 mg/mL | |
0.5 mL | |
Wet Ice | |
Streptavidin Conjugate (fluorescent) |
AP | |
Store in refrigerator (2–8°C). | |
Liquid | |
AP (Alkaline Phosphatase) |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Molecular Probes™ Streptavidin, Alkaline Phosphatase Conjugate > 0.5mL
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction