missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novex™ Streptavidin - HRP
Streptavidin - HRP
Brand: Novex™ 434323
290.32 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Reactivity: HRP-SA reacts with biotin.Applications: Immunohistochemistry, Western blotting, and ELISA
Specifications
1.25 mg/mL | |
2.5 mg | |
Wet Ice | |
Streptavidin Conjugate (fluorescent) |
HRP | |
Store in refrigerator (2–8°C). | |
Liquid | |
HRP (Horseradish Peroxidase) |
For Research Use Only. Not for use in diagnostic procedures.