missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Streptavidin Protein, HRP

Brand:  Thermo Scientific™ 21127

780.90 EUR valid until 2024-12-31
Use promo code "21979" to get your promotional price.


 View more versions of this product

Product Code. 10197673

  • 822.00 € / 5mg
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Streptavidin is biotin-binding protein that was originally isolated from Streptomyces avidinii. In contrast to avidin, streptavidin has no carbohydrate and has a mildly acidic pI of 5. Invitrogen Streptavidin products use a recombinant form of streptavidin having a mass of 53,000 daltons and a near-neutral pI. Streptavidin is a tetrameric protein, with each subunit binding one molecule of biotin with affinity similar to that of avidin. Guanidinium chloride will dissociate avidin and streptavidin into subunits, but streptavidin is more resistant to dissociation.
Formulation: 5mg purified protein in PBS and no preservative; pH 6.8
This product has been successfully used in ELISA, Immunohistochemistry and Western Blot procedures.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HRP
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, but do not store above -20°C.
Solid
Streptavidin Conjugate
HRP (Horseradish Peroxidase)
5 mg
Ambient Temperature
Pierce™
Biotin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.