Learn More
Invitrogen™ Streptavidin Protein, Rhodamine
Protein
Brand:  Invitrogen™ 21724
Product Code. 10697324
  • 330.00 € / 1mg
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Streptavidin Protein, Rhodamine conjugate, 21724, from Invitrogen., 21724, from Invitrogen. Species Reactivity: ; Applications: ELISA, Immunohistochemistry, Western Blot.
Streptavidin is biotin-binding protein that was originally isolated from Streptomyces avidinii. In contrast to avidin, streptavidin has no carbohydrate and has a mildly acidic pI of 5. Thermo Scientific Streptavidin products use a recombinant form of streptavidin having a mass of 53,000 daltons and a near-neutral pI. Streptavidin is a tetrameric protein, with each subunit binding one molecule of biotin with affinity similar to that of avidin. Guanidinium chloride will dissociate avidin and streptavidin into subunits, but streptavidin is more resistant to dissociation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rhodamine
PBS with no preservative
RUO
Lyophilized
purified
Streptavidin
Tag
1mg
4°C
ELISA, Immunohistochemistry, Western Blot
N/A