Learn More
Streptozocin, Tocris Bioscience™
DNA alkylator; antitumor and induces diabetes
110.00 €
Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18762714
View Documents
Tocris
1621/100
100 mg
110.00 €
100mg
Description
Description
Antibiotic and antitumor agent. Alkylates DNA and induces diabetes mellitus via reduction of nicotinamide adenine dinucleotide in pancreatic β-cells in vivo.
Videos
Safety and Handling
Safety and Handling
Recommended Storage : Store at -20°C