missing translation for 'onlineSavingsMsg'
Learn More

Cytiva StrepTrap™ HP Prepacked Columns

Purify Strep(II)-tagged recombinant proteins by affinity chromatography. Cytiva Life Sciences™ StrepTrap™ HP Prepacked Columns are packed with a robust and stable Sepharose™ based medium which results in high resolution, sharp concentrated peaks, and reproducible purifications. The binding capacity is also high.

171.65 € - 1994.33 €

Specifications

Product Type StrepTactin Sepharose HP prepacked column
Height (English) 0.98 in.
Height (Metric) 25mm
Material Polypropylene
Ligand Type StrepTactin (∼5mg/mL medium)
View More Specs

Products 4
Product Code Brand Quantity Flow Rate Content And Storage Price Quantity & Availability  
Product Code Brand Quantity Flow Rate Content And Storage Price Quantity & Availability  
10298754
View Documents Promotion Details
Cytiva
28-9075-47
1 x 5mL <20mL/min. 4° to 8°C This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11540654
View Documents Promotion Details
Cytiva
28-9075-48
5 x 5mL <20mL/min. 4° to 8°C This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10288754
View Documents Promotion Details
Cytiva
28-9075-46
5 x 1mL <4mL/min. 2° to 8°C This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15117956
View Documents Promotion Details
Cytiva
29048653
1 x 1mL <4mL/min. 2° to 8°C This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Prepacked with StrepTactin Sepharose High Performance
  • Binding capacity Approx. 6 mg Strep II-tagged protein/ml medium
  • Maximum pressure: 3 bar
  • Column material: polypropylene
  • Compatible with a wide range of reducing agents, detergents, denaturants, and other additives
  • Generated with 0.5M NaOH
  • 1 and 5mL columns

Media
  • Ligand: Streptactin
  • Matrix: 6% cross-linked agarose
  • Average particle size: 34μm
  • pH stability working range: >7

1mL columns

  • Bed dimensions: 7 x 25mm
  • Flow velocity: <4mL/min.

5mL columns
  • Bed dimensions: 16 x 25mm
  • Flow velocity: <20mL/min.

Specifications

Specifications

StrepTactin Sepharose HP prepacked column
25mm
StrepTactin (∼5mg/mL medium)
7
6% Highly Cross-linked Agarose
Purification of Strep (II) -tagged recombinant Proteins by affinity chromato graphy
0.98 in.
Polypropylene
3 bar
34μm
Ion Exchange Column
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • StrepTrap HP, 5 ml, 1 x 5 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Product Certifications