missing translation for 'onlineSavingsMsg'
Learn More

Cytiva StrepTrap XT Chromatography Column

StrepTrap XP is a column packed with Strep-Tactin™ XT Sepharose resin to purify Strep-tag™ II and Twin-Strep-tag™ recombinant proteins.

Brand:  Cytiva 29401320

 View more versions of this product

Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 16456149

  • 597.00 € / Pack of 5
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For use with Strep-Tactin™ XT Sepharose™
  • Prepacked in chromatography columns for convenience
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 mL
5 x 1 mL
1 mL/min.
Purification of recombinant proteins tagged with Strep-tag™ II and Twin-Strep-tag™
7 mm
25 mm
Affinity Column
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers