missing translation for 'onlineSavingsMsg'
Learn More

Cytiva StrepTrap XT Chromatography Column

StrepTrap XP is a column packed with Strep-Tactin™ XT Sepharose resin to purify Strep-tag™ II and Twin-Strep-tag™ recombinant proteins.

Brand:  Cytiva 29401323


 View more versions of this product

Product Code. 16476149

  • 2465.00 € / Pack of 5
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For use with Strep-Tactin™ XT Sepharose™
  • Prepacked in chromatography columns for convenience
TRUSTED_SUSTAINABILITY
Specifications

Specifications

5 mL
5 x 5 mL
5 mL/min.
Purification of recombinant proteins tagged with Strep-tag™ II and Twin-Strep-tag™
16 mm
25 mm
Affinity Column
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • StrepTrap XT, 5 x 5 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers