missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ String Knits™ 76-100 Series Natural Cotton Light Weight Gloves

Can be worn on their own or as liners. Permits a longer, more comfortable wear with no weak points which might split or fray.

26.75 € - 27.82 €

Specifications

Back Material Cotton
Construction Seamless Knit
Cuff Material Cotton
Color Natural
Grade 10 gauge
View More Specs

Products 2
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
12696961
View Documents
Ansell™
76100070
7
26.75 €
12 pairs
In Stock
to see stock.
Edge
Add to basket
 
12606971
View Documents
Ansell™
76100090
9
27.82 €
12 pairs
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

  • Stringknit construction allows good air flow to the hand.
  • Hands don't become hot and sweaty.
  • Ambidextrous: No need to pair, therefore more economical.
  • All styles washable at 40°C.
  • Will not shrink, stiffen or lose their shape.
  • More hygienic for the wearer, better value for the provider.
Specifications

Specifications

Cotton
Cotton
10 gauge
76-100
Cotton
Mechanical protection
Seamless Knit
Natural
Cotton
Natural
12 Pairs
76-100
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell™ String Knits™ 76-100 Series Natural Cotton Light Weight Gloves

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.