missing translation for 'onlineSavingsMsg'
Learn More

Ansell Edmont™ String Knits™ 76-160 Series Natural Cotton Light Weight Gloves

Product Code. 12646971
Click to view available options
Size:
6
7
8
9
10
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 12646971

Brand: Ansell Edmont™ 76160060

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Can be worn on their own or as liners. Permits a longer, more comfortable wear with no weak points which might split or fray.

  • Stringknit construction allows good air flow to the hand.
  • Hands don't become hot and sweaty.
  • Ambidextrous: No need to pair, therefore more economical.
  • All styles washable at 40°C.
  • Will not shrink, stiffen or lose their shape.
  • More hygienic for the wearer, better value for the provider.
TRUSTED_SUSTAINABILITY

Specifications

Back Material Polyester/Cotton Blend
Construction Seamless Knit
Cuff Material Polyester/Cotton
Color Natural
Grade 13 gauge
Material Polyester, Cotton
Product Type Light Weight Gloves
Size 6
Style 76-160
Glove Cuff Color Natural
Glove Cuff Style Knitwrist
Lining Polyester/Cotton
Quantity 12 Pairs
Resistant To Mechanical protection
Series 76-160
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.