missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Stripettor™ Ultra Pipet Controller
GREENER_CHOICE

A light, ergonomic, motorized pipet controller designed for use with glass and plastic serological pipets from 0.5 to 100mL

Brand:  Corning™ 4099

286.16 EUR valid until 2024-11-29
Use promo code "21683" to get your promotional price.


Product Code. 15536304

  • 372.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Description

Description

Conveniently positioned switches change operating modes and speeds quickly to handle different liquid volumes and various viscosities. Aspirating and dispensing speed is controlled through the finger triggers. When fully charged, the batteries enable up to 8 hours of continuous use.
  • LCD display for continuous visualization of speed, modes, and battery status
  • Additional colored nose cones to customize by application or user
  • Quiet powerful motor
  • Well positioned function buttons
  • Includes charging station (4998)
TRUSTED_SUSTAINABILITY
Spécification

Spécification

AAA
LCD
Glass and plastic serological pipets from 0.5 to 100mL
2
Pipette Controller
0.2μm
Universal power supply, Charging station (4998), three 0.2μm filters, three colored pieces and wall hanger
1
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Product Certifications
Offres spéciales

Offres spéciales