Learn More
Corning™ Stripwell™ Microplates
GREENER_CHOICE
Designed and treated to provide uniform binding, high optical clarity, and low background interference. Corning™ Stripwell™ Microplates provide a superior solution for in vitro diagnostic assays.
Brand:  Corning™ 2592
562.51 EUR valid until 2024-12-31
Use promo code "22007" to get your promotional price.
 View more versions of this product
Product Code. 10517691
  • 623.83 € / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Corning™ Stripwell™ microplates may be used in a variety of laboratory assays. The flat bottom strips are designed to easily break apart and are pre-assembled in an “egg-crate” style strip holder that allows each individual well to be positioned back into the microplate once broken.
  • Stripwell microplates have 96-well flat bottom polystyrene format.
  • Low volume and standard Stripwell microplates have well volumes of 190 μL and 360 μL, respectively.
  • 1 x 8 strips are designed to fit only one way into the strip holder, eliminating the chance of misorientation.
Order Info
Bulk packed. Cat. No. 07-200-24 has individual 1 x 8 strips without frame.
Certifications
Cat. Nos. 07-200-31 and -32 have certified surface chemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
High Binding
Without Lid
High Bind
Polystyrene
75 to 200 μL
High Binding
Round, Flat Bottom (F-well)
100/Cs.
100/Cs.
Strip Plate
8 x 12
Clear
96
Non-sterile
360 μL
In-vitro diagnostic assays
25/Pk.