missing translation for 'onlineSavingsMsg'
Learn More

Strontium acetate hydrate, Puratronic™, 99.9965% (metals basis), Thermo Scientific Chemicals

Product Code. 11362088
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 543-94-2
Molecular Formula C4H6O4Sr
Molecular Weight (g/mol) 205.71
MDL Number MFCD00036392
InChI Key RXSHXLOMRZJCLB-UHFFFAOYSA-L
Synonym strontium acetate, strontium diacetate, acetic acid, strontium salt, strontium di acetate, unii-4sl32ymy7b, 4sl32ymy7b, acetic acid, strontium salt 2:1, acmc-1bn7n, strontium 2+ ion diacetate, ksc492a5d
PubChem CID 10987
SMILES [Sr++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 150°C (in -0.5H2O)
Physical Form Crystalline
Quantity 5 g
Linear Formula Sr(OOCCH3)2·xH2O
Beilstein 3692534
Merck Index 14,8836
IUPAC Name strontium;diacetate
Formula Weight 205.71 (Anhydrous)
Percent Purity 99.9965%
Grade Puratronic™
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Strontium acetate hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.