missing translation for 'onlineSavingsMsg'
Learn More

Strontium acetate hydrate, Puratronic™, 99.9965% (metals basis), Thermo Scientific Chemicals

104.52 € - 366.83 €

Chemical Identifiers

CAS 543-94-2
Molecular Formula C4H6O4Sr
Molecular Weight (g/mol) 205.71
MDL Number MFCD00036392
InChI Key RXSHXLOMRZJCLB-UHFFFAOYSA-L
Synonym strontium acetate, strontium diacetate, acetic acid, strontium salt, strontium di acetate, unii-4sl32ymy7b, 4sl32ymy7b, acetic acid, strontium salt 2:1, acmc-1bn7n, strontium 2+ ion diacetate, ksc492a5d
PubChem CID 10987
SMILES [Sr++].CC([O-])=O.CC([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11362088
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
012990.06
5 g
104.52 €
5g
Please to purchase this item. Need a web account? Register with us today!
11352088
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
012990.14
25 g
366.83 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

543-94-2
205.71
RXSHXLOMRZJCLB-UHFFFAOYSA-L
10987
C4H6O4Sr
MFCD00036392
strontium acetate, strontium diacetate, acetic acid, strontium salt, strontium di acetate, unii-4sl32ymy7b, 4sl32ymy7b, acetic acid, strontium salt 2:1, acmc-1bn7n, strontium 2+ ion diacetate, ksc492a5d
[Sr++].CC([O-])=O.CC([O-])=O

Specifications

150°C (in -0.5H2O)
Crystalline
Sr(OOCCH3)2·xH2O
3692534
strontium acetate, strontium diacetate, acetic acid, strontium salt, strontium di acetate, unii-4sl32ymy7b, 4sl32ymy7b, acetic acid, strontium salt 2:1, acmc-1bn7n, strontium 2+ ion diacetate, ksc492a5d
[Sr++].CC([O-])=O.CC([O-])=O
205.71
205.71 (Anhydrous)
Puratronic™
(metals basis)
543-94-2
C4H6O4Sr
MFCD00036392
14,8836
RXSHXLOMRZJCLB-UHFFFAOYSA-L
strontium;diacetate
10987
99.9965%
Hygroscopic
Strontium acetate hydrate
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 208-854-8

missing translation for 'rtecsNumber' : AJ4725000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only