missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Strontium aluminate, 99.5% (metals basis)
Click to view available options
Quantity:
25 g
100 g
Packungsgröße:
100g
25g
Beschreibung
Strontium aluminate acts as a photo luminescent phosphor with long persistence of phosphorescence.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
| Physical Form | Powder |
| Quantity | 25 g |
| Assay Percent Range | 99.5% (metals basis) |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 205.58 |
| Density | 3.559 |
| Chemical Name or Material | Strontium aluminate |
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur