missing translation for 'onlineSavingsMsg'
Learn More

Strontium bromide, anhydrous, 99% (metals basis excluding Ba), Ba 0.1% max, Thermo Scientific Chemicals

203.00 € - 768.00 €

Chemical Identifiers

CAS 10476-81-0
Molecular Formula Br2Sr
Molecular Weight (g/mol) 247.428
MDL Number MFCD00049548
InChI Key YJPVTCSBVRMESK-UHFFFAOYSA-L
Synonym acmc-20ajmt, strontium bromide srbr2, strontium 2+ ion dibromide
PubChem CID 5360351
IUPAC Name strontium;dibromide
SMILES [Br-].[Br-].[Sr+2]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11387208
View Documents Promotion Details
Thermo Scientific Alfa Aesar
016791.18
50 g
203.00 €
50g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11377208
View Documents Promotion Details
Thermo Scientific Alfa Aesar
016791.30
250 g
768.00 €
250g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Used as sedatives in medicine and analytical reagents, for facile synthesis of poly(phenylcarbyne), which is a precursor to diamond-like carbon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10476-81-0
247.428
YJPVTCSBVRMESK-UHFFFAOYSA-L
5360351
[Br-].[Br-].[Sr+2]
Br2Sr
MFCD00049548
acmc-20ajmt, strontium bromide srbr2, strontium 2+ ion dibromide
strontium;dibromide

Specifications

643°C
White
Br2Sr
MFCD00049548
acmc-20ajmt, strontium bromide srbr2, strontium 2+ ion dibromide
YJPVTCSBVRMESK-UHFFFAOYSA-L
strontium;dibromide
5360351
99%
Hygroscopic
(metals basis excluding Ba), Ba 0.1% max.
10476-81-0
Granules
SrBr2
14,8837
Soluble in water,carbinol,ethanol,amyl alcohol. Insoluble in ether.
[Br-].[Br-].[Sr+2]
247.428
247.44
Odorless
4.216 g/mL
Strontium bromide, Anhydrous
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 233-969-5

missing translation for 'rtecsNumber' : WK8050000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only