missing translation for 'onlineSavingsMsg'
Learn More

Strontium carbonate, Puratronic™, 99.994% (metals basis), Thermo Scientific Chemicals

Used as a colorant in fireworks. It is also used in ceramics industry.

Chemical Identifiers

CAS 1633-05-2
Molecular Formula CO3Sr
Molecular Weight (g/mol) 147.63
MDL Number MFCD00011250
InChI Key LEDMRZGFZIAGGB-UHFFFAOYSA-L
Synonym strontium carbonate, strontianite, carbonic acid, strontium salt 1:1, unii-41ypu4mmca, strontium carbonate srco3, ccris 3203, carbonic acid strontium salt 1:1, 41ypu4mmca, strontium, reference standard solution, strontiumcarbonate
PubChem CID 15407
IUPAC Name strontium;carbonate
SMILES [Sr++].[O-]C([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11388058
View Documents Promotion Details
Thermo Scientific Alfa Aesar
036618.14
25 g N/A
N/A
11368058
View Documents Promotion Details
Thermo Scientific Alfa Aesar
036618.22
100 g N/A
N/A
11378058
View Documents Promotion Details
Thermo Scientific Alfa Aesar
036618.36
500 g N/A
N/A
Description

Description

Used as a colorant in fireworks. It is also used in ceramics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1633-05-2
147.63
LEDMRZGFZIAGGB-UHFFFAOYSA-L
15407
[Sr++].[O-]C([O-])=O
CO3Sr
MFCD00011250
strontium carbonate, strontianite, carbonic acid, strontium salt 1:1, unii-41ypu4mmca, strontium carbonate srco3, ccris 3203, carbonic acid strontium salt 1:1, 41ypu4mmca, strontium, reference standard solution, strontiumcarbonate
strontium;carbonate

Specifications

1100°C (decomposition)
Powder
MFCD00011250
Soluble in ammonium chloride. Slightly soluble in ammonia and water.
[Sr++].[O-]C([O-])=O
147.63
147.63
Plastic bottle
3.7
1633-05-2
CO3Sr
strontium carbonate, strontianite, carbonic acid, strontium salt 1:1, unii-41ypu4mmca, strontium carbonate srco3, ccris 3203, carbonic acid strontium salt 1:1, 41ypu4mmca, strontium, reference standard solution, strontiumcarbonate
LEDMRZGFZIAGGB-UHFFFAOYSA-L
strontium;carbonate
15407
99.994%
3.7
Strontium carbonate
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 216-643-7

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only