missing translation for 'onlineSavingsMsg'
Learn More

Strontium chloride, 99.99%, (trace metal basis), anhydrous

Product Code. 10008961 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10008961

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10476-85-4 | Cl2Sr | 158.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10476-85-4
Molecular Formula Cl2Sr
Molecular Weight (g/mol) 158.53
MDL Number MFCD00011249

Specifications

Boiling Point 1250.0°C
Melting Point 873.0°C
Color White
Physical Form Crystalline Powder
Quantity 5 g
Assay Percent Range 54.4 to 55.9% (Sr)
Linear Formula SrCl2
Merck Index 15, 8970
Solubility Information Solubility in water: soluble
Formula Weight 158.53
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Strontium chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.