Learn More
Strontium chloride hexahydrate, ACS, 99.0-103.0%, Thermo Scientific Chemicals
Description
Strontium chloride hexahydrate is a precursor to prepare strontium chromate which is used as a corrosion inhibitor for aluminum. It is involved in the synthesis of cadmium sulfide core photocatalytic nanoparticles. It is used as a red coloring agent in pyrotechnics. Furthermore, it is utilized in glass-making and metallurgy. In the medical field, its radioactive isotope strontium-89 is used for the treatment of bone cancer. It acts as a catalyst in the conversion of ketones and aldehydes to the corresponding gem-dihydroperoxides using aqueous hydrogen peroxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Physical Form | Granules |
Quantity | 100 g |
Assay Percent Range | 99.0 to 103.0% |
Merck Index | 14,8840 |
Solubility Information | Soluble in water. Slightly soluble in ethanol and acetone. Insoluble in ammonia. |
Formula Weight | 266.62 (158.53 Anhydrous) |
Percent Purity | 99.0 to 103.0% |
Grade | ACS |
Refractive Index | 1.65 |
Sensitivity | Hygroscopic |
Show More |
Safety and Handling

- Strontium chloride hexahydrate
- Danger
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Strontium chloride hexahydrate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.