missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Strontium chloride hexahydrate, Puratronic™, 99.9965% (metals basis), Thermo Scientific Chemicals
88.00 € - 303.00 €
Chemical Identifiers
CAS | 10025-70-4 |
---|---|
Molecular Formula | Cl2H12O6Sr |
Molecular Weight (g/mol) | 266.61 |
MDL Number | MFCD00149865 |
InChI Key | AMGRXJSJSONEEG-UHFFFAOYSA-L |
IUPAC Name | strontium(2+) hexahydrate dichloride |
SMILES | O.O.O.O.O.O.[Cl-].[Cl-].[Sr++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11307047
|
Thermo Scientific Alfa Aesar
010877.06 |
5 g |
88.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11397037
|
Thermo Scientific Alfa Aesar
010877.14 |
25 g |
303.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10025-70-4 | |
266.61 | |
AMGRXJSJSONEEG-UHFFFAOYSA-L | |
O.O.O.O.O.O.[Cl-].[Cl-].[Sr++] |
Cl2H12O6Sr | |
MFCD00149865 | |
strontium(2+) hexahydrate dichloride |
Specifications
10025-70-4 | |
Powder | |
MFCD00149865 | |
AMGRXJSJSONEEG-UHFFFAOYSA-L | |
strontium(2+) hexahydrate dichloride | |
266.62 (158.53 Anhydrous) | |
Puratronic™ | |
Hygroscopic | |
(metals basis) |
White | |
Cl2H12O6Sr | |
14,8840 | |
O.O.O.O.O.O.[Cl-].[Cl-].[Sr++] | |
266.61 | |
99.9965% | |
1.65 | |
1.93 g/mL | |
Strontium chloride hexahydrate |
Safety and Handling
GHS H Statement
H318
Causes serious eye damage.
P261-P271-P280-P304+P340-P305+P351+P338-P310-P312-P501c
H318-H335
missing translation for 'einecsNumber' : 233-971-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only