missing translation for 'onlineSavingsMsg'
Learn More

Strontium chloride hexahydrate, Puriss., ≥99%, Honeywell Fluka™

Product Code. 15691370
Click to view available options
Quantity:
100 g
500 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Puriss.,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-70-4
Molecular Formula Cl2H12O6Sr
Molecular Weight (g/mol) 266.61
MDL Number MFCD00149865
InChI Key AMGRXJSJSONEEG-UHFFFAOYSA-L
IUPAC Name strontium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Sr++]

Specifications

Quantity 1 kg
Linear Formula SrCl2 · 6H2O
UN Number NONH for all modes of transport
Formula Weight 266.62g/mol
Percent Purity ≥99%
Grade puriss.
Chemical Name or Material Strontium Chloride Hexahydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'