missing translation for 'onlineSavingsMsg'
Learn More

Strontium chloride hexahydrate, tech., Thermo Scientific Chemicals

55.48 € - 154.77 €

Chemical Identifiers

CAS 10025-70-4
Molecular Formula Cl2H12O6Sr
Molecular Weight (g/mol) 266.61
MDL Number MFCD00149865
InChI Key AMGRXJSJSONEEG-UHFFFAOYSA-L
IUPAC Name strontium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Sr++]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11381829
SDS View Documents
Thermo Scientific Alfa Aesar
012494.36
500 g
55.48 €
500g
Please to purchase this item. Need a web account? Register with us today!
11391829
SDS View Documents
Thermo Scientific Alfa Aesar
012494.A3
2 kg
154.77 €
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Strontium chloride hexahydrate is a precursor to prepare strontium chromate which is used as a corrosion inhibitor for aluminum. It is involved in the synthesis of cadmium sulfide core photocatalytic nanoparticles. It is used as a red coloring agent in pyrotechnics. Furthermore, it is utilized in glass-making and metallurgy. In the field of medicine, its radioactive isotope strontium-89 is used for the treatment of bone cancer. It acts as a catalyst in the conversion of ketones and aldehydes to the corresponding gem-dihydroperoxides using aqueous hydrogen peroxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10025-70-4
266.61
AMGRXJSJSONEEG-UHFFFAOYSA-L
O.O.O.O.O.O.[Cl-].[Cl-].[Sr++]
Cl2H12O6Sr
MFCD00149865
strontium(2+) hexahydrate dichloride

Specifications

10025-70-4
Cl2H12O6Sr
Soluble in water,alcohol. Slightly soluble in ethanol and acetone. Insoluble in ammonia.
O.O.O.O.O.O.[Cl-].[Cl-].[Sr++]
266.61
Technical
Hygroscopic
Strontium chloride hexahydrate
Crystalline
MFCD00149865
AMGRXJSJSONEEG-UHFFFAOYSA-L
strontium(2+) hexahydrate dichloride
266.62 (158.53 Anhydrous)
1.65
1.93 g/mL
Videos
Safety and Handling

Safety and Handling

P261-P271-P280-P304+P340-P305+P351+P338-P310-P312-P501c

H318-H335

missing translation for 'einecsNumber' : 233-971-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Strontium chloride hexahydrate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H318-Causes serious eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Strontium chloride hexahydrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.