missing translation for 'onlineSavingsMsg'
Learn More

Strontium chromate, 95%, Thermo Scientific Chemicals

81.71 € - 243.21 €

Chemical Identifiers

CAS 7789-06-2
Molecular Formula CrO4Sr
Molecular Weight (g/mol) 203.61
MDL Number MFCD00049549
InChI Key NVKTUNLPFJHLCG-UHFFFAOYSA-N
Synonym strontium chromate, strontium yellow, strontium chromate vi, sutokuro t, deep lemon yellow, strontium chromate a, c.i. pigment yellow 32, strontium chromate 1:1, strontium chromate 12170
PubChem CID 24599
IUPAC Name strontium;dioxido(dioxo)chromium
SMILES [O-][Cr](=O)(=O)[O-].[Sr+2]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15462987
SDS View Documents
Thermo Scientific Alfa Aesar
089026.30
250 g
81.71 €
250g
Please to purchase this item. Need a web account? Register with us today!
15472987
SDS View Documents
Thermo Scientific Alfa Aesar
089026.A1
1 kg
243.21 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Strontium chromate is used an primer for zinc, magnesium, aluminum, and alloys used in aircraft manufacture. Used as a colorant in polyvinyl chloride resins. It is also used in aluminum flake coatings. paints, varnishes and oil colors. It is a used in water based wash primers, metal conditioners, additive to control the sulfate content of solutions in electrochemical processes. It is also used in paints.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7789-06-2
203.61
NVKTUNLPFJHLCG-UHFFFAOYSA-N
24599
[O-][Cr](=O)(=O)[O-].[Sr+2]
CrO4Sr
MFCD00049549
strontium chromate, strontium yellow, strontium chromate vi, sutokuro t, deep lemon yellow, strontium chromate a, c.i. pigment yellow 32, strontium chromate 1:1, strontium chromate 12170
strontium;dioxido(dioxo)chromium

Specifications

7789-06-2
CrO4Sr
strontium chromate, strontium yellow, strontium chromate vi, sutokuro t, deep lemon yellow, strontium chromate a, c.i. pigment yellow 32, strontium chromate 1:1, strontium chromate 12170
NVKTUNLPFJHLCG-UHFFFAOYSA-N
strontium;dioxido(dioxo)chromium
24599
3.895
Powder
MFCD00049549
Soluble in boiling water,dilute HCl,dilute CH3CO2H
[O-][Cr](=O)(=O)[O-].[Sr+2]
203.61
203.61
Strontium chromate
Videos
Safety and Handling

Safety and Handling

P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P284-P301+P312-P302+P352-P304+P340-P308+P313-P310-P330-P333+P313-P363-P501c

H302-H317-H330-H335-H340-H350-H361

missing translation for 'dotInformation' : Hazard Class: 9; Packaging Group: III

missing translation for 'einecsNumber' : 232-142-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.