missing translation for 'onlineSavingsMsg'
Learn More

Strontium granules, 20mm max, 99.0% (metals basis), Thermo Scientific Chemicals

2310.50 €

Chemical Identifiers

CAS 7440-24-6
Molecular Formula Sr
Molecular Weight (g/mol) 87.62
MDL Number MFCD00134060
InChI Key CIOAGBVUUVVLOB-UHFFFAOYSA-N
Synonym strontium, elemental, unii-yzs2rpe8le, yzs2rpe8le, estroncio, granules, 19mm 0.76in & down metals basis, strontium, distilled dendritic pieces, atom, pieces, strontium, chunks, pieces, dendritic
PubChem CID 5359327
ChEBI CHEBI:33324
IUPAC Name strontium
SMILES [Sr]
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11320198
SDS View Documents
Thermo Scientific Alfa Aesar
042929.14
25 g
2310.50 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
Specifications

Chemical Identifiers

7440-24-6
87.62
CIOAGBVUUVVLOB-UHFFFAOYSA-N
5359327
strontium
Sr
MFCD00134060
strontium, elemental, unii-yzs2rpe8le, yzs2rpe8le, estroncio, granules, 19mm 0.76in & down metals basis, strontium, distilled dendritic pieces, atom, pieces, strontium, chunks, pieces, dendritic
CHEBI:33324
[Sr]

Specifications

Golden
Sr
UN3208
CIOAGBVUUVVLOB-UHFFFAOYSA-N
strontium
5359327
99.8%
Air and moisture sensitive
Strontium, Distilled dendritic pieces
7440-24-6
MFCD00134060
strontium, elemental, unii-yzs2rpe8le, yzs2rpe8le, estroncio, granules, 19mm 0.76in & down metals basis, strontium, distilled dendritic pieces, atom, pieces, strontium, chunks, pieces, dendritic
[Sr]
87.62
CHEBI:33324
Ampule
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H261
In contact with water releases flammable gas.

P223-P231+P232-P280-P335+P334-P370+P378q-P501c

H260

missing translation for 'dotInformation' : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: METALLIC SUBSTANCE, WATER-REACTIVE, N.O.S.

missing translation for 'einecsNumber' : 231-133-4

missing translation for 'rtecsNumber' : WK7700000

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Ampouled under argon

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.