Learn More
Strontium dodecairon nonadecaoxide, 99.0% (metals basis), Thermo Scientific Chemicals
Description
Strontium dodecairon nonadecaoxide is used in manufacture of mixtures, granulates for plastic articles, production of magnet articles by compacting, sintering or other processes. And it is also used as an additive to flares, pyrotechnics, provides high refractive index to glass.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | >450°C |
Physical Form | Powder |
Quantity | 5 g |
Solubility Information | Soluble in organic solvents. |
Formula Weight | 1061.77 |
Percent Purity | 99.5% |
Assay | (metals basis) |
Chemical Name or Material | Strontium dodecairon nonadecaoxide |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.