missing translation for 'onlineSavingsMsg'
Learn More

Strontium iodide, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11395637
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Strontium iodide is used as a scintillation gamma radiation detector. It is also used in a range of hand-held radiation detection instruments as well as in medical, industrial and environmental applications. It is used in medicine as a substitute for potassium iodide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10476-86-5
Molecular Formula SrI2
MDL Number MFCD00049557

Specifications

Melting Point 515°C
Color White to Yellow
Physical Form Powder
Quantity 100 g
Merck Index 14,8844
Solubility Information Soluble in water and ethanol.
Formula Weight 341.43
Percent Purity 99.99%
Odor Odorless
Sensitivity Hygroscopic; Light sensitive
Density 4.550 g/mL
Assay (metals basis)
Chemical Name or Material Strontium iodide, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'