Learn More
Strontium nitrate, anhydrous, 99.97% (metals basis), Thermo Scientific Chemicals
121.00 € - 358.00 €
Chemical Identifiers
CAS | 10042-76-9 |
---|---|
Molecular Formula | N2O6Sr |
Molecular Weight (g/mol) | 211.63 |
MDL Number | MFCD00011248 |
InChI Key | DHEQXMRUPNDRPG-UHFFFAOYSA-N |
Synonym | strontium nitrate, strontium dinitrate, nitric acid, strontium salt, strontium ii nitrate 1:2, unii-bdg873aqzl, nitrate de strontium french, hsdb 787, strontium nitrate sr no3 2, bdg873aqzl, nitric acid, strontium salt 2:1 |
PubChem CID | 24848 |
SMILES | [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11347288
|
Thermo Scientific Alfa Aesar
030486.14 |
25 g |
121.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11337288
|
Thermo Scientific Alfa Aesar
030486.22 |
100 g |
358.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Strontium nitrate is used in the production of road flares and in fireworks. It serves as a colorant (red) in pyrotechnics and is also used as an oxidizer in pyrotechnics. It mixed with glycolic acid and used to reduce the sensation of skin irritation. It is also used in electrophysiology experiments.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10042-76-9 | |
211.63 | |
DHEQXMRUPNDRPG-UHFFFAOYSA-N | |
24848 |
N2O6Sr | |
MFCD00011248 | |
strontium nitrate, strontium dinitrate, nitric acid, strontium salt, strontium ii nitrate 1:2, unii-bdg873aqzl, nitrate de strontium french, hsdb 787, strontium nitrate sr no3 2, bdg873aqzl, nitric acid, strontium salt 2:1 | |
[Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
570°C | |
White | |
N2O6Sr | |
MFCD00011248 | |
14,8845 | |
Soluble in water and ammonia. Slightly soluble in alcohol,ethanol and acetone. Insoluble in nitric acid. | |
[Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
211.63 | |
211.63 | |
Odorless | |
2.990 g/mL | |
Strontium nitrate, Anhydrous |
10042-76-9 | |
Crystalline | |
Sr(NO3)2 | |
UN1507 | |
strontium nitrate, strontium dinitrate, nitric acid, strontium salt, strontium ii nitrate 1:2, unii-bdg873aqzl, nitrate de strontium french, hsdb 787, strontium nitrate sr no3 2, bdg873aqzl, nitric acid, strontium salt 2:1 | |
DHEQXMRUPNDRPG-UHFFFAOYSA-N | |
strontium(2+) dinitrate | |
24848 | |
99.97% | |
Hygroscopic | |
(metals basis) |
Safety and Handling
GHS H Statement
H272-H302-H315-H319-H335
May intensify fire
oxidizer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P220-P221-P280-P305+P351+P338-P310-P370+P378q-P501c
H272-H318
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: STRONTIUM NITRATE
missing translation for 'einecsNumber' : 233-131-9
missing translation for 'rtecsNumber' : WK9800000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Oxidising solid Category 1
Hazard Statement
- H271-May cause fire or explosion; strong oxidiser.
- H318-Causes serious eye damage.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
RUO – Research Use Only