missing translation for 'onlineSavingsMsg'
Learn More

Strontium oxide, 99.5% (metals basis), SrO ^=97%, Thermo Scientific Chemicals

548.00 €

Chemical Identifiers

CAS 1314-11-0
Molecular Formula OSr
Molecular Weight (g/mol) 103.619
MDL Number MFCD00049558
InChI Key UFQXGXDIJMBKTC-UHFFFAOYSA-N
PubChem CID 518709
IUPAC Name oxostrontium
SMILES O=[Sr]
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11339219
SDS View Documents
Thermo Scientific Alfa Aesar
088220.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Strontium oxide was used to make television picture tube glass. It finds application in glass, optic and ceramic industry. It is an active component of paint dryers. It is also used to prepare pure strontium up on heating with aluminum under vacuum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1314-11-0
103.619
UFQXGXDIJMBKTC-UHFFFAOYSA-N
oxostrontium
OSr
MFCD00049558
518709
O=[Sr]

Specifications

ca 3000°C
1314-11-0
99.5% (metals basis)
MFCD00049558
UFQXGXDIJMBKTC-UHFFFAOYSA-N
oxostrontium
518709
Moisture sensitive
Strontium oxide
2430°C
Powder
OSr
Soluble in potassium hydroxide solution. Slightly soluble in alcohol. Insoluble in acetone and ether.
O=[Sr]
103.619
103.62
4.7 g/mL
Videos
Safety and Handling

Safety and Handling

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II

missing translation for 'einecsNumber' : 215-219-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Strontium oxide

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Strontium oxide (SrO)

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.