missing translation for 'onlineSavingsMsg'
Learn More

Strontium oxide, tech., Thermo Scientific Chemicals

Product Code. 11321839
Click to view available options
Quantity:
250 g
1 kg
5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

In the manufacture of strontium salts, pyrotechnics, pigments, in greases and soaps and as a desiccant.Strontium oxide was used to make television picture tube glass. It finds application in glass, optic and ceramic industries. It is an active component of paint dryers. It is also used to prepare pure strontium upon heating with aluminum under vacuum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • In the manufacture of strontium salts, pyrotechnics, and pigments, in greases and soaps, and as a desiccant
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-11-0
Molecular Formula OSr
Molecular Weight (g/mol) 103.619
MDL Number MFCD00049558
InChI Key UFQXGXDIJMBKTC-UHFFFAOYSA-N
PubChem CID 518709
IUPAC Name oxostrontium
SMILES O=[Sr]

Specifications

Boiling Point ∼3000°C
Melting Point 2430°C
Physical Form Powder
Quantity 250 g
UN Number UN3262
Merck Index 14,8846
Solubility Information Soluble in fused potassium hydroxide; Insoluble in ether and acetone; Decomposes in water to Sr(OH)2 with evolution of heat;Slightly soluble in alcohol; Insoluble in acetone and ether.
Formula Weight 103.62
Grade Technical
Odor Odorless
Sensitivity Moisture sensitive
Density 4.700 g/mL
Chemical Name or Material Strontium oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.