missing translation for 'onlineSavingsMsg'
Learn More

Strontium perchlorate trihydrate, 98%, Thermo Scientific Chemicals

Product Code. 15472377
Click to view available options
Quantity:
10 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Strontium perchlorate trihydrate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15650-09-6
Molecular Formula Cl2H6O11Sr
Molecular Weight (g/mol) 340.56
MDL Number MFCD09039117
InChI Key VRRVWIQWZWFYDJ-UHFFFAOYSA-L
Synonym strontium perchlorate trihydrate, 2clo4.sr.3h2o, strontium perchlorate, trihydrate, strontium 2+ trihydrate diperchlorate
PubChem CID 71435311
SMILES O.O.O.[Sr++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O

Specifications

Physical Form Crystalline
Quantity 10 g
Assay Percent Range 98%
Solubility Information Very soluble in water. Soluble in alcohol.
IUPAC Name strontium;diperchlorate;trihydrate
Formula Weight 340.54 (286.52 Anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Strontium perchlorate trihydrate
compliance-icons compliance-icons
Product Identifier
  • Strontium perchlorate trihydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Strontium perchlorate trihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.