Learn More
Strontium perchlorate trihydrate, 98%, Thermo Scientific Chemicals
26.13 € - 180.90 €
Chemical Identifiers
CAS | 15650-09-6 |
---|---|
Molecular Formula | Cl2H6O11Sr |
Molecular Weight (g/mol) | 340.56 |
MDL Number | MFCD09039117 |
InChI Key | VRRVWIQWZWFYDJ-UHFFFAOYSA-L |
Synonym | strontium perchlorate trihydrate, 2clo4.sr.3h2o, strontium perchlorate, trihydrate, strontium 2+ trihydrate diperchlorate |
PubChem CID | 71435311 |
SMILES | O.O.O.[Sr++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15472377
|
Thermo Scientific Alfa Aesar
045577.09 |
10 g |
26.13 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15482377
|
Thermo Scientific Alfa Aesar
045577.22 |
100 g |
56.98 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15492377
|
Thermo Scientific Alfa Aesar
045577.36 |
500 g |
180.90 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Strontium perchlorate trihydrate is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15650-09-6 | |
340.56 | |
VRRVWIQWZWFYDJ-UHFFFAOYSA-L | |
71435311 |
Cl2H6O11Sr | |
MFCD09039117 | |
strontium perchlorate trihydrate, 2clo4.sr.3h2o, strontium perchlorate, trihydrate, strontium 2+ trihydrate diperchlorate | |
O.O.O.[Sr++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O |
Specifications
15650-09-6 | |
98% | |
MFCD09039117 | |
Very soluble in water. Soluble in alcohol. | |
O.O.O.[Sr++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O | |
340.56 | |
340.54 (286.52 Anhydrous) | |
Strontium perchlorate trihydrate |
Crystalline | |
Cl2H6O11Sr | |
strontium perchlorate trihydrate, 2clo4.sr.3h2o, strontium perchlorate, trihydrate, strontium 2+ trihydrate diperchlorate | |
VRRVWIQWZWFYDJ-UHFFFAOYSA-L | |
strontium;diperchlorate;trihydrate | |
71435311 | |
Hygroscopic |
Safety and Handling
P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H272-H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 5.1; Packaging Group: II
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Strontium perchlorate trihydrate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash face, hands, and anyexposed skin thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Strontium perchlorate trihydrate
RUO – Research Use Only