missing translation for 'onlineSavingsMsg'
Learn More

Strontium Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical

Product Code. 10538430
Click to view available options
Quantity:
500 mL
Packaging:
HDPE plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Packaging HDPE plastic bottle
Quantity 500 mL
Color Colorless
MDL Number 134060
pH 1
Physical Form Liquid
compliance-icons
Product Identifier
  • Strontium solution 1000 ppm
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.