missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Strontium sulfate, Puratronic™, 99.9965% (metals basis), Thermo Scientific Chemicals
90.40 € - 318.00 €
Chemical Identifiers
CAS | 7759-02-6 |
---|---|
Molecular Formula | O4SSr |
Molecular Weight (g/mol) | 183.68 |
MDL Number | MFCD00036274 |
InChI Key | UBXAKNTVXQMEAG-UHFFFAOYSA-L |
Synonym | strontium sulfate, strontium sulphate, sulfuric acid, strontium salt 1:1, strontium 2+ ion sulfate, unii-7q3kx2l47f, strontiumsulfat, celestine, strontium 2+ sulfate, strontium sulfate 25g |
PubChem CID | 3084026 |
IUPAC Name | strontium;sulfate |
SMILES | [O-]S(=O)(=O)[O-].[Sr+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11397597
|
Thermo Scientific Alfa Aesar
012870.06 |
5 g |
90.40 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11387597
|
Thermo Scientific Alfa Aesar
012870.14 |
25 g |
318.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7759-02-6 | |
183.68 | |
UBXAKNTVXQMEAG-UHFFFAOYSA-L | |
3084026 | |
[O-]S(=O)(=O)[O-].[Sr+2] |
O4SSr | |
MFCD00036274 | |
strontium sulfate, strontium sulphate, sulfuric acid, strontium salt 1:1, strontium 2+ ion sulfate, unii-7q3kx2l47f, strontiumsulfat, celestine, strontium 2+ sulfate, strontium sulfate 25g | |
strontium;sulfate |
Specifications
1600°C | |
White | |
O4SSr | |
MFCD00036274 | |
strontium sulfate, strontium sulphate, sulfuric acid, strontium salt 1:1, strontium 2+ ion sulfate, unii-7q3kx2l47f, strontiumsulfat, celestine, strontium 2+ sulfate, strontium sulfate 25g | |
[O-]S(=O)(=O)[O-].[Sr+2] | |
183.68 | |
183.68 | |
Puratronic™ | |
3.960 g/mL | |
Strontium sulfate |
7759-02-6 | |
Powder | |
SrSO4 | |
14,8848 | |
UBXAKNTVXQMEAG-UHFFFAOYSA-L | |
strontium;sulfate | |
3084026 | |
99.9965% | |
Odorless | |
(metals basis) |
Safety and Handling
missing translation for 'einecsNumber' : 231-850-2
missing translation for 'rtecsNumber' : WT1210000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only