Learn More
Strontium titanium oxide, S.A. 12.5-25m2/g, Thermo Scientific Chemicals
192.00 € - 722.00 €
Chemical Identifiers
CAS | 12060-59-2 |
---|---|
Molecular Formula | O3SrTi |
Molecular Weight (g/mol) | 183.48 |
MDL Number | MFCD00049554 |
InChI Key | VEALVRVVWBQVSL-UHFFFAOYSA-N |
Synonym | strontium titanate, strontium titanium oxide, unii-olh4i98373, o3ti.sr, strontium titanium oxide srtio3, strontium titanium trioxide, titanate tio32-, strontium 1:1, strontium titanate 100g, strontium dioxido oxo titanium, strontium titanate, powder |
PubChem CID | 82899 |
IUPAC Name | strontium;dioxido(oxo)titanium |
SMILES | [Sr++].[O-][Ti]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11378768
|
Thermo Scientific Alfa Aesar
041634.22 |
100 g |
192.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11388768
|
Thermo Scientific Alfa Aesar
041634.36 |
500 g |
722.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Strontium titaniumoxide is used as paint . It has been used as a strontium-90-containing material in radioisotope thermoelectric generators. It is also used as a diamond simulant. This compound is been used in television sets, radios and computers, capacitors, because of the dielectric constant of the material. Used in electronics and in electrical insulation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12060-59-2 | |
183.48 | |
VEALVRVVWBQVSL-UHFFFAOYSA-N | |
82899 | |
[Sr++].[O-][Ti]([O-])=O |
O3SrTi | |
MFCD00049554 | |
strontium titanate, strontium titanium oxide, unii-olh4i98373, o3ti.sr, strontium titanium oxide srtio3, strontium titanium trioxide, titanate tio32-, strontium 1:1, strontium titanate 100g, strontium dioxido oxo titanium, strontium titanate, powder | |
strontium;dioxido(oxo)titanium |
Specifications
2060°C | |
Powder | |
MFCD00049554 | |
It is insoluble in water and most solvents. | |
[Sr++].[O-][Ti]([O-])=O | |
183.48 | |
183.51 | |
4.810 g/mL |
12060-59-2 | |
O3SrTi | |
strontium titanate, strontium titanium oxide, unii-olh4i98373, o3ti.sr, strontium titanium oxide srtio3, strontium titanium trioxide, titanate tio32-, strontium 1:1, strontium titanate 100g, strontium dioxido oxo titanium, strontium titanate, powder | |
VEALVRVVWBQVSL-UHFFFAOYSA-N | |
strontium;dioxido(oxo)titanium | |
82899 | |
Odorless | |
Strontium titanium oxide |
Safety and Handling
missing translation for 'einecsNumber' : 235-044-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only