missing translation for 'onlineSavingsMsg'
Learn More

Strontium Titanium Oxide Substrate, 6.25 x 6.25 x 0.5mm, Polished One Side, 110 Orientation, Thermo Scientific™

Product Code. 11389228
Click to view available options
Quantity:
1 Ea.
5 Ea.
This item is not returnable. View return policy
This item is not returnable. View return policy

100 Orientation

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12060-59-2
Molecular Formula O3SrTi
Molecular Weight (g/mol) 183.48
MDL Number MFCD00049554
InChI Key VEALVRVVWBQVSL-UHFFFAOYSA-N
Synonym strontium titanate, strontium titanium oxide, unii-olh4i98373, o3ti.sr, strontium titanium oxide srtio3, strontium titanium trioxide, titanate tio32-, strontium 1:1, strontium titanate 100g, strontium dioxido oxo titanium, strontium titanate, powder
PubChem CID 82899
IUPAC Name strontium;dioxido(oxo)titanium
SMILES [Sr++].[O-][Ti]([O-])=O

Specifications

Color White
Physical Form Substrate
Quantity 1 Ea.
Odor Odorless
Chemical Name or Material Strontium Titanium Oxide Substrate, Polished One Side
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'