missing translation for 'onlineSavingsMsg'
Learn More

Strontium zirconium oxide, 99.3% (metals basis), Thermo Scientific Chemicals

Product Code. 11311719
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Mostly as intermediates( primary and secondary ). For chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12036-39-4
Molecular Formula O3SrZr
Molecular Weight (g/mol) 226.84
MDL Number MFCD00049555
InChI Key FCCTVDGKMTZSPU-UHFFFAOYSA-N
Synonym strontium zirconate, strontium zirconium trioxide, strontium dioxido oxo zirconium, o3zr.sr, strontium zirconium oxide srzro3, strontium zirconium oxide, strontium 2+ ion oxozirconiumbis olate, strontium 2+ oxozirconiumbis olate
PubChem CID 82852
IUPAC Name strontium;dioxido(oxo)zirconium
SMILES [Sr++].[O-][Zr]([O-])=O

Specifications

Boiling Point 9000°C
Melting Point 2700°C
Color White
Physical Form Powder
Quantity 1 kg
Formula Weight 226.84
Percent Purity 99.3%
Odor Odorless
Density 5.5 g/mL
Assay (metals basis)
Chemical Name or Material Strontium zirconium oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'