Learn More
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11479878
|
Thermo Scientific™
DR0585A |
Streptococcal Grouping Kit |
400.00 €
50 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11903522
|
Thermo Scientific™
DR0586G |
Streptococcal Grouping Reagent A |
114.00 €
50 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11913522
|
Thermo Scientific™
DR0587G |
Streptococcal Grouping Reagent B |
114.00 €
50 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10537283
|
Thermo Scientific™
DR0588G |
Streptococcal Grouping Reagent C |
114.00 €
50 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11973932
|
Thermo Scientific™
DR0589G |
Streptococcal Grouping Reagent D |
114.00 €
50 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10770354
|
Thermo Scientific™
DR0590G |
Streptococcal Grouping Reagent F |
114.00 €
50 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10127942
|
Thermo Scientific™
DR0591G |
Streptococcal Grouping Reagent G |
114.00 €
50 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10035812
|
Thermo Scientific™
DR0592G |
Polyvalent Positive Control; 50 tests |
121.00 €
50 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13275599
|
Thermo Scientific™
DR0593G |
Streptococcal Grouping Enzyme Extraction Reagent |
105.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rapidly identify streptococcal groups A, B, C, D, F, and G by latex agglutination using Thermo Scientific™ Oxoid™ Streptococcal Grouping Kit. This rapid latex agglutination test is designed to deliver simple, rapid and sensitive results for the identification of streptococcal groups A, B, C, D, F, and G . The test utilizes latex particles coated with group-specific antibodies which will agglutinate in the presence of homologous antigens.
Streptococci are facultatively anaerobic, catalase-negative, Gram-positive cocci that occur as pairs or chains. They cause a wide variety of clinical diseases including pharyngitis, scarlet fever, otitis media, impetigo, meningitis, neonatal sepis, pneumonia, endocarditis, osteomyelitis, and necrotizing fasciitis.
Use this rapid, simple and sensitive test for the identification of streptococcal groups A, B, C, D, F, and G.
- Rapid—results in two minutes
- Simple—easy to implement
- Visual—easy-to-read results
- Sensitive—use of latex technology makes this test more sensitive
- All reagents available separately
Lancefield showed that the majority of pathogenic streptococci possess specific carbohydrate antigens, which permit the classification of streptococci into groups. These streptococcal group antigens can be extracted from the cells and their presence demonstrated with latex particles previously coated with group-specific antibodies. These latex particles will agglutinate in the presence of homologous antigen, but will remain in smooth suspension in the absence of such antigen. The Oxoid Streptococcal Grouping Kit is such a latex agglutination test for the identification of the streptococcal groups and reagents are provided for groups A, B, C, D, F and G. The use of a enzymatic extraction procedure considerably shortens the time required for antigen extraction and much improves the antigen yield, partially for Group D streptococci.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Safety and Handling
- STREPTOCOCCAL GROUPING KIT
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H301-Toxic if swallowed.
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H373-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
- H311-Toxic in contact with skin.
- H332-Harmful if inhaled.
Precautionary Statement
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
- MIXTURE LIST-Contains : Achromopeptidase, Thiomersal, Sodium azide
Your input is important to us. Please complete this form to provide feedback related to the content on this product.