Learn More
Cole-Parmer™ Block Heat Insert for Cole-Parmer™ BH-200 and BH-250 Series Block Heaters and CB-200 Series Chiller/Heater
Interchangeable heating blocks offer excellent thermal conductivity.
172.00 € - 515.00 €
Product Code | Brand | Description | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Holds | Price | Quantity & Availability | |||||
11593809
|
Cole-Parmer™
36620-61 |
Blank with Thermometer Hole | Tubes, Plates |
192.00 €
Each |
Estimated Shipment: 20-05-2025
Log in to see stock. |
|||||
11936498
|
Cole-Parmer™
36610-50 |
Single Wide | 12 Tubes, 17 x 47 mm (dia. x L) |
172.00 €
Each |
In Stock
Log in to see stock. |
|||||
11476588
|
Cole-Parmer™
36610-63 |
Single Wide | 80 x 0.2 mL Tube Strips |
233.00 €
Each |
Estimated Shipment: 20-05-2025
Log in to see stock. |
|||||
12942590
|
Cole-Parmer™
36620-75 |
Gelation Timer Cup, Flat Bottom | Tubes, Plates |
515.00 €
Each |
Estimated Shipment: 16-07-2025
Log in to see stock. |
|||||
15556413
|
Cole-Parmer™
36620-92 |
Flat Bottom Wells | 20 x 13 mm dia. Vials |
192.00 €
Each |
Estimated Shipment: 20-05-2025
Log in to see stock. |
|||||
12071602
|
Cole-Parmer™
36620-65 |
Tapered Wells | 20 x 1.5 mL Tubes |
224.00 €
Each |
In Stock
Log in to see stock. |
|||||
12081602
|
Cole-Parmer™
36620-66 |
Tapered Wells | 30 x 0.5 mL Tubes |
192.00 €
Each |
In Stock
Log in to see stock. |
|||||
12952590
|
Cole-Parmer™
36620-82 |
Shallow, Conical | 20 x 12 mm Vials |
300.00 €
Each |
Estimated Shipment: 05-08-2025
Log in to see stock. |
|||||
15294497
|
Cole-Parmer™
36620-77 |
Conical Wells | 5 x 50 mL Tubes |
239.00 €
Pack of 5 |
In Stock
Log in to see stock. |
|||||
15319943
|
Cole-Parmer™
36620-79 |
Conical Wells | 5 x 30 mm dia. Tubes |
192.00 €
Each |
In Stock
Log in to see stock. |
|||||
11869564
|
Cole-Parmer™
36620-63 |
Conical Wells | 6 x 24 mm dia. Tubes/Vials |
192.00 €
Each |
Estimated Shipment: 20-05-2025
Log in to see stock. |
|||||
10129190
|
Cole-Parmer™
36620-64 |
Conical Wells | 6 x 26 mm dia. Tubes/Vials |
210.00 €
Each |
In Stock
Log in to see stock. |
|||||
11543809
|
Cole-Parmer™
36620-56 |
Conical Wells | 12 x 16 mm dia. Tubes/Vials |
255.84 €
Each |
In Stock
Log in to see stock. |
|||||
15218043
|
Cole-Parmer™
36620-81 |
Conical Wells | 8 x 20 mm dia. Tubes |
192.00 €
Each |
In Stock
Log in to see stock. |
|||||
12001612
|
Cole-Parmer™
36620-71 |
Conical Wells | 20 x 2 mL Tubes |
224.00 €
Each |
In Stock
Log in to see stock. |
|||||
11880705
|
Cole-Parmer™
36620-52 |
Conical Wells | 20 x 10 mm dia. Tubes/Vials |
192.00 €
Each |
In Stock
Log in to see stock. |
|||||
11523809
|
Cole-Parmer™
36620-54 |
Conical Wells | 20 x 13 mm dia. Tubes/Vials |
192.00 €
Each |
Estimated Shipment: 16-05-2025
Log in to see stock. |
|||||
11899694
|
Cole-Parmer™
36620-51 |
Conical Wells | 30 x 6 mm dia. Tubes/Vials |
192.00 €
Each |
Estimated Shipment: 02-09-2025
Log in to see stock. |
|||||
Description
- Versatile—interchangeable heating blocks accommodate various tubes and plates
- Block heaters sold separately
Interchangeable aluminum heating blocks provide excellent thermal conductivity and accommodate various test tubes, cups, and plates. Depending on the combination of blocks selected, plastic spacers (36620-67) can be used to fill a half-width recess, ensuring optimal thermal performance. Most blocks come with a separate hole to accept a thermometer.
Order a flexi-block to accommodate multiple size tubes, vials, or cuvettes at once. Unskirted PCR plates can be heated in either size flexi-block. Skirted microtiter plates, deep-well plates, and PCR plates can only be incubated in the triple-width version.
Specifications
Anodized Aluminum | |
Heating Block |